Assay Details
| Gene Symbol: vtd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015132.2 | NP_001015132.1 | 6 - 7 | 1943 | 141 | |
| GenBank mRNA | AF109926.1 | - | 6 - 7 | 1893 | 141 | |
| BT100037.1 | - | 6 - 7 | 1890 | 141 | ||
More Information
Set Membership: |
|

vtd
Genomic Map