Assay Details
| Gene Symbol: MED21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015223.3 | NP_001015223.1 | 1 - 2 | 180 | 85 | |
| NM_001015224.3 | NP_001015224.1 | 2 - 3 | 184 | 85 | ||
| GenBank mRNA | AY071027.1 | - | 2 - 3 | 93 | 85 | |
| KX532046.1 | - | 1 - 2 | 45 | 85 | ||
More Information
Set Membership: |
|

MED21
Genomic Map