Assay Details
| Gene Symbol: Rbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104161.3 | NP_001097631.1 | 4 - 5 | 1214 | 100 | |
| NM_001275050.2 | NP_001261979.1 | 4 - 5 | 1214 | 100 | ||
| NM_168718.3 | NP_730245.2 | 4 - 5 | 1214 | 100 | ||
| GenBank mRNA | AY122226.1 | - | 4 - 5 | 1212 | 100 | |
More Information
Set Membership: |
|

Rbp6
Genomic Map