Assay Details
| Gene Symbol: CG17691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015354.4 | NP_001015354.3 | 1 - 2 | 319 | 67 | |
| NM_001110546.2 | NP_001104016.2 | 1 - 2 | 319 | 67 | ||
| NM_001110548.2 | NP_001104018.1 | 2 - 3 | 342 | 67 | ||
More Information
Set Membership: |
|

CG17691
Genomic Map