Assay Details
| Gene Symbol: CG17167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015256.3 | NP_001015256.2 | 1 - 2 | 372 | 100 | |
| GenBank mRNA | BT099540.1 | - | 1 - 2 | 341 | 100 | |
More Information
Set Membership: |
|

CG17167
Genomic Map