Assay Details
| Gene Symbol: CG17167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015256.3 | NP_001015256.2 | 6 - 7 | 1545 | 70 | |
| NM_001015257.3 | NP_001015257.2 | 6 - 7 | 1548 | 70 | ||
| NM_001300717.1 | NP_001287646.1 | 6 - 7 | 1260 | 70 | ||
| GenBank mRNA | BT099540.1 | - | 6 - 7 | 1514 | 70 | |
| BT125892.1 | - | 6 - 7 | 1545 | 70 | ||
More Information
Set Membership: |
|

CG17167
Genomic Map