Assay Details
| Gene Symbol: CG17167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015257.3 | NP_001015257.2 | 1 - 2 | 382 | 103 | |
| NM_001300716.1 | NP_001287645.1 | 1 - 2 | 382 | 103 | ||
| GenBank mRNA | BT125892.1 | - | 1 - 2 | 379 | 103 | |
More Information
Set Membership: |
|

CG17167
Genomic Map