Assay Details
Gene Symbol: CG12567 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015384.3 | NP_001015384.1 | 1 - 2 | 330 | 82 | |
NM_001110861.3 | NP_001104331.1 | 1 - 2 | 330 | 82 | ||
NM_001170363.2 | NP_001163834.1 | 1 - 2 | 330 | 82 | ||
NM_001300704.1 | NP_001287633.1 | 1 - 2 | 93 | 82 | ||
GenBank mRNA | BT083411.1 | - | 1 - 2 | 325 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
