Assay Details
| Gene Symbol: UQCR-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170381.2 | NP_001163852.1 | 1 - 2 | 73 | 82 | |
| NM_001300710.1 | NP_001287639.1 | 1 - 2 | 73 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001170381.2, NM_001300710.1 transcripts.
Set Membership: |
|

UQCR-11
Genomic Map