Assay Details
| Gene Symbol: Parp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110982.3 | NP_001104452.1 | 2 - 3 | 326 | 110 | |
| GenBank mRNA | BT015238.1 | - | 2 - 3 | 324 | 110 | |
| D13806.1 | - | 2 - 3 | 293 | 110 | ||
More Information
Set Membership: |
|

Parp
Genomic Map