Assay Details
| Gene Symbol: Parp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110982.3 | NP_001104452.1 | 4 - 5 | 1192 | 168 | |
| GenBank mRNA | BT015238.1 | - | 4 - 5 | 1190 | 168 | |
| D13806.1 | - | 4 - 5 | 1159 | 168 | ||
More Information
Set Membership: |
|

Parp
Genomic Map