Assay Details
| Gene Symbol: nht | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078850.4 | NP_523574.4 | 1 | 644 | 66 | |
| GenBank mRNA | BT022194.1 | - | 1 | 643 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nht| Gene Symbol: nht | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078850.4 | NP_523574.4 | 1 | 644 | 66 | |
| GenBank mRNA | BT022194.1 | - | 1 | 643 | 66 | |
Set Membership: |
|