Assay Details
| Gene Symbol: SkpF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137952.3 | NP_611796.1 | 1 | 151 | 67 | |
| GenBank mRNA | AY118610.1 | - | 1 | 148 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SkpF| Gene Symbol: SkpF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_137952.3 | NP_611796.1 | 1 | 151 | 67 | |
| GenBank mRNA | AY118610.1 | - | 1 | 148 | 67 | |
Set Membership: |
|