Assay Details
| Gene Symbol: Haspin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015349.4 | NP_001015349.2 | 1 - 2 | 137 | 113 | |
| GenBank mRNA | AY118897.1 | - | 1 - 2 | 165 | 113 | |
More Information
Set Membership: |
|

Haspin
Genomic Map