Assay Details
| Gene Symbol: Scp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015389.3 | NP_001015389.1 | 2 - 3 | 131 | 87 | |
| NM_001276231.1 | NP_001263160.1 | 2 - 3 | 172 | 87 | ||
| GenBank mRNA | AY118788.1 | - | 1 - 2 | 109 | 87 | |
More Information
Set Membership: |
|

Scp1
Genomic Map