Assay Details
| Gene Symbol: Scp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015389.3 | NP_001015389.1 | 5 - 6 | 533 | 135 | |
| NM_001276231.1 | NP_001263160.1 | 5 - 6 | 574 | 135 | ||
| GenBank mRNA | AY118788.1 | - | 4 - 5 | 511 | 135 | |
More Information
Set Membership: |
|

Scp1
Genomic Map