Assay Details
| Gene Symbol: lt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042950.3 | NP_001036415.2 | 6 - 7 | 1226 | 111 | |
| NM_001042951.2 | NP_001036416.1 | 6 - 7 | 1226 | 111 | ||
| NM_001042952.2 | NP_001036417.1 | 5 - 6 | 1278 | 111 | ||
| NM_001273766.1 | NP_001260695.1 | 6 - 7 | 1226 | 111 | ||
| GenBank mRNA | AY118388.1 | - | 5 - 6 | 1286 | 111 | |
| BT024228.1 | - | 6 - 7 | 1183 | 111 | ||
More Information
Set Membership: |
|

lt
Genomic Map