Assay Details
| Gene Symbol: cta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042956.2 | NP_001036421.1 | 1 - 2 | 779 | 95 | |
| NM_001273767.1 | NP_001260696.1 | 1 - 2 | 779 | 95 | ||
| GenBank mRNA | AY089534.1 | - | 1 - 2 | 760 | 95 | |
| M94285.1 | - | 1 - 2 | 530 | 95 | ||
More Information
Set Membership: |
|

cta
Genomic Map