Assay Details
| Gene Symbol: Spf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042961.3 | NP_001036426.1 | 1 - 2 | 192 | 76 | |
| GenBank mRNA | BT044104.1 | - | 1 - 2 | 253 | 76 | |
| DQ295864.1 | - | 1 - 2 | 162 | 76 | ||
More Information
Set Membership: |
|

Spf45
Genomic Map