Assay Details
| Gene Symbol: crabp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001842.1 | NP_001001842.1 | 2 - 3 | 355 | 87 | |
| GenBank mRNA | AY616861.1 | - | 2 - 3 | 355 | 87 | |
| BC162180.1 | - | 2 - 3 | 332 | 87 | ||
| BC162190.1 | - | 2 - 3 | 332 | 87 | ||
More Information
Set Membership: |
|

crabp1b
Genomic Map