Assay Details
| Gene Symbol: srsf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002053.2 | NP_001002053.1 | 4 - 5 | 521 | 94 | |
| NM_001328122.2 | NP_001315051.1 | 5 - 6 | 527 | 94 | ||
| XM_009302539.3 | XP_009300814.1 | 5 - 6 | 635 | 94 | ||
| GenBank mRNA | BC071322.1 | - | 4 - 5 | 521 | 94 | |
| EH527237.1 | - | 3 - 4 | 488 | 94 | ||
| EH564513.1 | - | 3 - 4 | 488 | 94 | ||
| EH606039.1 | - | 3 - 4 | 494 | 94 | ||
More Information
Set Membership: |
|

srsf3a
Genomic Map