Assay Details
| Gene Symbol: srsf3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002053.2 | NP_001002053.1 | 6 - 7 | 641 | 115 | |
| NM_001328122.2 | NP_001315051.1 | 7 - 8 | 647 | 115 | ||
| XM_009302539.3 | XP_009300814.1 | 7 - 8 | 755 | 115 | ||
| GenBank mRNA | BC071322.1 | - | 6 - 7 | 641 | 115 | |
| EH518653.1 | - | 1 - 2 | 396 | 115 | ||
| EH527237.1 | - | 1 - 2 | 368 | 115 | ||
| EH564513.1 | - | 1 - 2 | 368 | 115 | ||
More Information
Set Membership: |
|

srsf3a
Genomic Map