Assay Details
| Gene Symbol: pa2g4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002070.2 | NP_001002070.1 | 11 - 12 | 1110 | 90 | |
| GenBank mRNA | BC071407.1 | - | 11 - 12 | 1110 | 90 | |
More Information
Set Membership: |
|

pa2g4a
Genomic Map