Assay Details
| Gene Symbol: kdm5bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002166.2 | NP_001002166.2 | 19 - 20 | 3091 | 93 | |
| XM_005167024.4 | XP_005167081.1 | 20 - 21 | 3769 | 93 | ||
| XM_009303989.3 | XP_009302264.1 | 20 - 21 | 3769 | 93 | ||
| GenBank mRNA | BC071280.1 | - | 19 - 20 | 3024 | 93 | |
More Information
Set Membership: |
|

kdm5bb
Genomic Map