Assay Details
| Gene Symbol: srprb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002572.1 | NP_001002572.1 | 6 - 7 | 619 | 61 | |
| GenBank mRNA | BC076224.1 | - | 6 - 7 | 619 | 61 | |
| BC095560.1 | - | 6 - 7 | 615 | 61 | ||
| BC164607.1 | - | 6 - 7 | 624 | 61 | ||
More Information
Set Membership: |
|

srprb
Genomic Map