Assay Details
| Gene Symbol: rab30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002750.2 | NP_001002750.1 | 4 - 5 | 538 | 67 | |
| GenBank mRNA | BC076437.1 | - | 4 - 5 | 542 | 67 | |
More Information
Set Membership: |
|

rab30
Genomic Map