Assay Details
| Gene Symbol: chchd4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003538.2 | NP_001003538.1 | 2 - 3 | 237 | 83 | |
| GenBank mRNA | BC077126.1 | - | 2 - 3 | 242 | 83 | |
More Information
Set Membership: |
|

chchd4a
Genomic Map