Assay Details
| Gene Symbol: cenpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003555.3 | NP_001003555.2 | 6 - 7 | 645 | 73 | |
| GenBank mRNA | BC078335.1 | - | 6 - 7 | 619 | 73 | |
More Information
Set Membership: |
|

cenpp
Genomic Map