Assay Details
| Gene Symbol: aurka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003640.2 | NP_001003640.1 | 6 - 7 | 926 | 61 | |
| GenBank mRNA | BC078304.1 | - | 6 - 7 | 930 | 61 | |
| KC136847.1 | - | 5 - 6 | 719 | 61 | ||
More Information
Set Membership: |
|

aurka
Genomic Map