Assay Details
| Gene Symbol: ptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003981.2 | NP_001003981.2 | 2 - 3 | 252 | 66 | |
| GenBank mRNA | AY572239.1 | - | 2 - 3 | 183 | 66 | |
| BC095549.1 | - | 2 - 3 | 252 | 66 | ||
| BC165895.1 | - | 1 - 2 | 119 | 66 | ||
| KM655586.1 | - | 1 - 2 | 85 | 66 | ||
More Information
Set Membership: |
|

ptn
Genomic Map