Assay Details
| Gene Symbol: ptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003981.2 | NP_001003981.2 | 4 - 5 | 582 | 56 | |
| GenBank mRNA | AY572239.1 | - | 4 - 5 | 513 | 56 | |
| BC095549.1 | - | 4 - 5 | 582 | 56 | ||
| BC165895.1 | - | 3 - 4 | 449 | 56 | ||
| KM655586.1 | - | 3 - 4 | 415 | 56 | ||
More Information
Set Membership: |
|

ptn
Genomic Map