Assay Details
| Gene Symbol: msna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004296.1 | NP_001004296.1 | 11 - 12 | 1397 | 71 | |
| XM_009301281.3 | XP_009299556.1 | 11 - 12 | 1295 | 71 | ||
| XM_009301282.3 | XP_009299557.1 | 7 - 8 | 840 | 71 | ||
| GenBank mRNA | BC049479.1 | - | 11 - 12 | 1461 | 71 | |
| BC081551.1 | - | 11 - 12 | 1397 | 71 | ||
More Information
Set Membership: |
|

msna
Genomic Map