Assay Details
| Gene Symbol: msna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004296.1 | NP_001004296.1 | 6 - 7 | 699 | 83 | |
| XM_009301281.3 | XP_009299556.1 | 6 - 7 | 597 | 83 | ||
| XM_009301282.3 | XP_009299557.1 | 2 - 3 | 142 | 83 | ||
| GenBank mRNA | BC049479.1 | - | 6 - 7 | 763 | 83 | |
| BC081551.1 | - | 6 - 7 | 699 | 83 | ||
More Information
Set Membership: |
|

msna
Genomic Map