Assay Details
| Gene Symbol: mpdu1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004545.1 | NP_001004545.1 | 3 - 4 | 405 | 82 | |
| GenBank mRNA | BC081598.1 | - | 3 - 4 | 405 | 82 | |
More Information
Set Membership: |
|

mpdu1a
Genomic Map