Assay Details
Gene Symbol: tmem237b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004636.1 | NP_001004636.1 | 7 - 8 | 539 | 105 | |
XM_009302154.3 | XP_009300429.1 | 6 - 7 | 469 | 105 | ||
GenBank mRNA | BC081384.1 | - | 7 - 8 | 539 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
