Assay Details
| Gene Symbol: arfip2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005935.1 | NP_001005935.1 | 3 - 4 | 369 | 89 | |
| XM_005157358.4 | XP_005157415.1 | 4 - 5 | 503 | 89 | ||
| GenBank mRNA | BC083521.1 | - | 3 - 4 | 369 | 89 | |
More Information
Set Membership: |
|

arfip2a
Genomic Map