Assay Details
| Gene Symbol: pttg1ipb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005941.1 | NP_001005941.1 | 2 - 3 | 229 | 124 | |
| XM_005167479.4 | XP_005167536.1 | 2 - 3 | 241 | 124 | ||
| GenBank mRNA | BC083469.1 | - | 2 - 3 | 229 | 124 | |
| BC164885.1 | - | 2 - 3 | 183 | 124 | ||
| KM655647.1 | - | 2 - 3 | 149 | 124 | ||
More Information
Set Membership: |
|

pttg1ipb
Genomic Map