Assay Details
| Gene Symbol: osr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006079.1 | NP_001006079.1 | 2 - 3 | 940 | 112 | |
| GenBank mRNA | BC083241.1 | - | 2 - 3 | 940 | 112 | |
| BC164601.1 | - | 1 - 2 | 695 | 112 | ||
More Information
Set Membership: |
|

osr1
Genomic Map