Assay Details
| Gene Symbol: cldn10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007037.1 | NP_001007038.1 | 1 - 2 | 219 | 66 | |
| GenBank mRNA | BC163751.1 | - | 1 - 2 | 289 | 66 | |
More Information
Set Membership: |
|

cldn10a
Genomic Map