Assay Details
| Gene Symbol: trmt10c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007332.2 | NP_001007333.2 | 3 - 4 | 740 | 79 | |
| GenBank mRNA | BC085556.1 | - | 3 - 4 | 369 | 79 | |
More Information
Set Membership: |
|

trmt10c
Genomic Map