Assay Details
| Gene Symbol: mapk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013451.3 | NP_001013469.2 | 6 - 7 | 2308 | 72 | |
| XM_005156002.4 | XP_005156059.1 | 5 - 6 | 3094 | 72 | ||
| GenBank mRNA | DQ360072.1 | - | 6 - 7 | 2308 | 72 | |
More Information
Set Membership: |
|

mapk7
Genomic Map