Assay Details
| Gene Symbol: spcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013469.1 | NP_001013487.1 | 4 - 5 | 440 | 92 | |
| GenBank mRNA | BC091463.1 | - | 4 - 5 | 440 | 92 | |
More Information
Set Membership: |
|

spcs2
Genomic Map