Assay Details
| Gene Symbol: mat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013474.2 | NP_001013492.2 | 5 - 6 | 790 | 99 | |
| XM_005161395.4 | XP_005161452.1 | 5 - 6 | 970 | 99 | ||
| GenBank mRNA | BC091455.1 | - | 5 - 6 | 728 | 99 | |
| EH549300.1 | - | 5 - 6 | 814 | 99 | ||
| EH566280.1 | - | 2 - 3 | 542 | 99 | ||
| EH580990.1 | - | 5 - 6 | 782 | 99 | ||
More Information
Set Membership: |
|

mat2b
Genomic Map