Assay Details
| Gene Symbol: ippk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014324.1 | NP_001014346.1 | 11 - 12 | 1264 | 61 | |
| GenBank mRNA | BC091783.1 | - | 11 - 12 | 1264 | 61 | |
| BC165752.1 | - | 11 - 12 | 1198 | 61 | ||
| DQ075212.1 | - | 11 - 12 | 1164 | 61 | ||
More Information
Set Membership: |
|

ippk
Genomic Map