Assay Details
| Gene Symbol: mrps25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017590.1 | NP_001017590.1 | 3 - 4 | 421 | 101 | |
| GenBank mRNA | BC092991.1 | - | 3 - 4 | 421 | 101 | |
More Information
Set Membership: |
|

mrps25
Genomic Map