Assay Details
| Gene Symbol: paplnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020564.1 | NP_001018400.1 | 11 - 12 | 1227 | 87 | |
| GenBank mRNA | BC095166.1 | - | 11 - 12 | 1227 | 87 | |
| BC165723.1 | - | 10 - 11 | 1207 | 87 | ||
More Information
Set Membership: |
|

paplnb
Genomic Map