Assay Details
| Gene Symbol: paplnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020564.1 | NP_001018400.1 | 13 - 14 | 1441 | 66 | |
| XM_005160691.4 | XP_005160748.1 | 13 - 14 | 1787 | 66 | ||
| GenBank mRNA | BC095166.1 | - | 13 - 14 | 1441 | 66 | |
| BC165723.1 | - | 12 - 13 | 1421 | 66 | ||
More Information
Set Membership: |
|

paplnb
Genomic Map