Assay Details
| Gene Symbol: paplnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001020564.1 | NP_001018400.1 | 8 - 9 | 808 | 62 | |
| XM_005160691.4 | XP_005160748.1 | 8 - 9 | 1184 | 62 | ||
| GenBank mRNA | BC095166.1 | - | 8 - 9 | 808 | 62 | |
| BC165723.1 | - | 7 - 8 | 788 | 62 | ||
More Information
Set Membership: |
|

paplnb
Genomic Map