Assay Details
| Gene Symbol: chordc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025175.1 | NP_001020346.1 | 4 - 5 | 367 | 86 | |
| GenBank mRNA | BC097168.1 | - | 4 - 5 | 367 | 86 | |
More Information
Set Membership: |
|

chordc1a
Genomic Map