Assay Details
| Gene Symbol: chordc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025175.1 | NP_001020346.1 | 10 - 11 | 831 | 74 | |
| GenBank mRNA | BC097168.1 | - | 10 - 11 | 831 | 74 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				

chordc1a
Genomic Map